96-Well Plate Cover Foils, Zymo Research
Supplier: Zymo Research
C2007-12 C2007-2 C2007-24 C2007-4 C2007-6 C2007-8
76004-234EA 48.03 CAD
76004-234 76004-236 76005-216 76005-218 76005-220 76005-222
96-Well Plate Cover Foils, Zymo Research
Aluminium Foils
Pierceable aluminum foil for sealing 96-well plates and blocks.
  • Strong adhesive for tight seals
  • Ideal for cold storage
Dimensions: 82.6×132.6 mm
Order Now
SPECIFICATIONS
Environmentally Preferable