Buffer P2 (Green), Zymo Research
Supplier: Zymo Research
D4027-2-10
D4027-2-160
D4027-2-20
D4027-2-250
D4027-2-320
D4027-2-80
76020-552EA
22.34
CAD
76020-552
76020-554
76020-556
76020-558
76020-560
76020-562
Buffer P2 (Green), Zymo Research
Buffers
Assay Buffers
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...