Buffer P1 (Red), Zymo Research
Supplier: Zymo Research
D4027-1-10
D4027-1-160
D4027-1-20
D4027-1-320
D4027-1-500
D4027-1-80
76020-540EA
22.34
CAD
76020-540
76020-542
76020-544
76020-546
76020-548
76020-550
Buffer P1 (Red), Zymo Research
Buffers
Assay Buffers
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...