Gorodkowa Agar, Modified, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M983-500G M983-500G
CA95022-586EA 1270.32 CAD
CA95022-586 95022-586
Gorodkowa Agar, Modified, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For promoting sporulation of yeasts
  • Microbiological culture media
  • 500 g poly bottles