Silver nitrate ≥99.7%, crystals, BAKER ANALYZED® ACS
Supplier: VWR International
|
Danger
|
Synonyms:
Lunar caustic, Silver mononitrate, Nitric acid silver salt
Management of Change (MOC) category = N
CAJT3426-1
CAJT3426-4
3426-00
CAJT3426-1CS
68441.65
CAD
CAJT3426-1
CAJT3426-4
JT3426-0
Silver nitrate ≥99.7%, crystals, BAKER ANALYZED® ACS
Silver nitrate
Formula:
AgNO₃ MW: 169.87 g/mol Boiling Pt: 444 Melting Pt: 210 Density: 4.35 g/cm³ |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 UN: 1493 ADR: 5.1 II Merck Index: 12,08661 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (AgNO₃) | ≥99.7 % |
Clarity of Solution | Passes Test |
Free Acid | Passes Test |
Substances Not Precipitated by HCl | ≤0.01 % |
Sulfate (SO₄) | ≤0.002 % |
Lead (Pb) | ≤0.001 % |
Chloride (Cl) | ≤5 ppm |
Trace Impurities - Copper (Cu) | ≤2.000 ppm |
Trace Impurities - Iron (Fe) | ≤2.0 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...