Potassium chloride 2 M in aqueous solution, sterile

Supplier: TEKNOVA CA

P0335 P0330
CA100219-982EA 324.26 CAD
CA100219-982 CA100219-992
Potassium chloride 2 M in aqueous solution, sterile
Potassium chloride
Formula: KCl
MDL Number: MFCD00011360
CAS Number: 7447-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR