Actidione Agar Base without Actidione, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
M058-500G
M058-2.5KG
M058-5KG
CA95020-150EA
1304.91
CAD
CA95020-150
CA89222-354
CA89222-356
Actidione Agar Base without Actidione, HiMedia
Media for Microbiology
Prepared Media for Microbiology
Actidione agar base without actidione is used for the enumeration and detection of bacteria in specimens containing large number of yeasts and moulds.