VWR® Cell Lifter
Supplier: VWR
VWR®
CSC012023 CSC011023
76036-004CS 181.97 CAD
76036-004 76036-006
VWR® Cell Lifter
Cell Scrapers
Flat blade cell lifters are manufactured with high-grade polyethylene, which is tensile to ensure minimal cell damage during manual harvesting.
  • Sterilized by electron beam irradiation
  • Non-pyrogenic and Dnase-/RNase-free
  • Total length is 234 mm
VWR® cell lifters are available with two different blade styles, the 2.5 mm wide Narrow blade or the 9.0 mm wide J-Hook blade. Both options are designed to provide convenient operation with blade designs that minimize the potential for cell damage.
Certifications: ISO13485:2003 and ISO9001:2008.