MacConkey Broth, Double Strength, with Neutral Red, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M539-500G M539S-500G
CA95021-964EA 973.11 CAD
CA95021-964 CA95021-966
MacConkey Broth, Double Strength, with Neutral Red, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the primary isolation of coliforms from large samples such as water or waste water.
  • Microbiological culture media
  • 500 g poly bottles