Hugh Leifson Medium Dehydrated Culture Media, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M826-500G M826S-500G
CA95022-314EA 1149.47 CAD
CA95022-314 CA95022-316
Hugh Leifson Medium Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For detecting aerobic and anaerobic breakdown of glucose.
  • Microbiological culture media
  • 500 g poly bottles