SP Wilmad-LabGlass Calibration/Measuring Flask Set, TC Metric System, SP Industries
Supplier: Avantor
14200-580EA 579.01 CAD
14200-580 14200-712 14200-736 14201-078 14201-086 14201-220 14212-738
SP Wilmad-LabGlass Calibration/Measuring Flask Set, TC Metric System, SP Industries
Flasks Volumetric Flasks
Borosilicate glass cylinder and flasks are calibrated to contain (TC) at 68 °F/20 °C.
  • The 50 ml is cylindrical, graduated 10 to 50 ml in 1 ml divisions
  • All others are graduated neck flasks, graduated above and below the nominal
Ordering information: The complete set includes a case which holds all six sizes.