Ethyl Violet Azide Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M426-500G M426S-500G
CA95021-808EA 952.2 CAD
CA95021-808 CA95021-810
Ethyl Violet Azide Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective confirmatory detection of enterococci as an indicator of fecal pollution in water and other specimens.
  • Microbiological culture media
  • 500 g poly bottles