Cell-Mate3D™ Cell Culture Kit, BRTI Life Sciences
Supplier: BRTI LIFE SCIENCES
CM-1002
CM-1001
76036-000EA
719.23
CAD
76036-000
76036-028
Cell-Mate3D™ Cell Culture Kit, BRTI Life Sciences
Assays
Cellular Assays
Defined tissue-like injectable 3D matrix used to grow tumors, stem cell organoids
- Chemically defined 3D cell culture
- Tissue-like matrix
- Injectable matrix
- Translational matrix (from research to clinical)
- Scalable multi-well plates
- 2 year shelf-life of kit
It is also used in tissue engineering and cancer research.