Lipovitellin Salt Mannitol Agar Base Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M627-500G M627-500G
CA95022-102EA 1344.74 CAD
CA95022-102 95022-102
Lipovitellin Salt Mannitol Agar Base Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and identification of pathogenic staphylococci by detecting lipase production and mannitol fermentation.
  • Microbiological culture media
  • 500 g poly bottles