Campylobacter Agar Base Dehydrated Culture Media, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
M994-500G
CA95022-608EA 1027.58 CAD
CA95022-608
Campylobacter Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of Campylobacter species from fecal specimens, food, and environmental specimens.
  • Microbiological culture media
  • 500 g poly bottles