Buffered Peptone Water, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M614-2.5KG M614-500G M614S-500G M614-10KG M614-25KG
CA89130-344EA 2776.98 CAD
CA89130-344 CA95022-076 CA95022-078 CA95059-662 CA71002-740
Buffered Peptone Water, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
A pre-enrichment medium for increasing the recovery of injured Salmonella species from foods prior to selective enrichment and isolation.
  • Microbiological culture media
  • Packed in poly bottles
Order Now
SPECIFICATIONS
Environmentally Preferable