PumpMaster Container Pumps, Burkle
Supplier: BURKLE INC
5202-3000 5202-2000 5202-1000
76047-104EA 602.49 CAD
76047-104 76047-108 76047-112
PumpMaster Container Pumps, Burkle
Pumps Drum/Transfer Pumps
The manual pressure pump is ideally suited for drawing off, decanting and emptying canisters and barrels.
  • Complete with three bungs in different sizes
The hand pump is used to create excess pressure that forces the liquid out of the container. The stopcock also allows extraction of small quantities of liquid. The PumpMaster container pump is mounted on the container using a flexible stopper with a variable and adjustable diameter. No special thread adapters are required. PumpMaster fits all commercial container openings with a diameter from 46.5 mm to 60 mm (1.83 to 2.36"). The immersion tube has a length of 95 cm (37.40") and it can be shortened depending on the size of the container.
Packaging: One pump per package.