Boyd Elite Crusher, Rocklabs
Supplier: ROCKLABS (W/T) CA
305873 305879 305876
CA76047-480EA 143325.26 CAD
CA76047-480 CA76047-484 CA76047-486
Boyd Elite Crusher, Rocklabs
Mills
The Boyd Elite allows the lab to get up to a 90% pass. No other crusher can consistently achieve this pass rate. The Boyd Elite accept lump sizes up to 70mm, an increase in input size of 40% which allows the lab to process larger samples through the crusher. The Boyd Elite incorporates improved dust and chip control, full electrial and mechanical safety and improved ergonomics.
  • Designed for trouble free operation and requires virtually no maintenance
  • The redesign results in virtualy no downtime
Featuring top and bottom moving jaws, the patented design of the Boyd Elite delivers the maximum amount of crushing, reducing samples by 35 times the original size
The Boyd Crusher and optional Rotating Sample Divider (RSD) have been designed to be used together for a one step crushing and splitting process. The sample is loaded into the Boyd Crusher then in one operation, it is crushed, conveyed to the RSD and then split into preset proportions of your choice.
Servicing is simple and can be undertaken after training by laboratory sample staff or trained service technicians. The Boyd Elite has been redesigned for trouble free operation, requires virtually no maintenance and is backed by a 3000 hour or 18 month warranty. Many areas have been redesigned to significantly reduce the wear and tear of the components and reduce the total cost of ownership.
Characterized by ease of maintenance and cleaning and equipped with stringent safety mechanisms as standard, the Boyd Elite reduces the time and cost associated with the crushing process.
Ordering information: The Boyd Elite is available with or without the Rotating Sample Divider (RSD). Manufacturer's 3000 hour or 18 month warranty (whichever comes first).