Easygel™ Nutrient Culture Media, Microbiology Laboratories
Supplier: MICROLOGY INC
NUT30 NUT40 NUT10
470111-890PK 181.7 CAD
470111-890 470193-386 470193-396
Easygel™ Nutrient Culture Media, Microbiology Laboratories
Media for Microbiology Dehydrated Media for Microbiology
This convenient, one-step media starts as a liquid and gels when poured into the provided petri dishes.
  • Pectin-Gel Method
  • Sterile
  • Pre-Mixed
  • No Autoclaving
  • No Condensation
Easygel is a pectin-gel method. As with traditional agar methods, you can add your sample to the liquid medium to create pour plates or let the gel harden to make streak plates.
No heating is needed, which reduces preparation time in the lab and makes it easy to take samples on field trips.
Ordering information: These products contain living or perishable materials. Easygel comes in a variety of types. The pre-treated petri dishes measure 100 x 15 mm. Shelf life is six months.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.