Decarboxylase Test Medium Base, Dehydrated, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M912-500G M912S-500G
CA95022-450EA 1112.86 CAD
CA95022-450 CA95022-452
Decarboxylase Test Medium Base, Dehydrated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For testing amino acid decarboxylase activity.
  • Microbiological culture media
  • 500 g poly bottles