Columbia Blood Agar Base Dehydrated Culture Media, HiMedia

Supplier: HIMEDIA LABORATORIES PVT. CA

M144-2.5KG M144-500G M144-5KG
CA89132-898EA 3947.25 CAD
CA89132-898 CA95021-134 CA89224-036
Columbia Blood Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology

An efficient base for the preparation of blood agar or chocolate agar, and for various selective and identification media and isolation of organisms from clinical and non clinical samples.


  • Cream to yellow homogeneous free flowing powder
  • Firm gelling, comparable with 1.5% Agar gel
  • pH: 7.10 to 7.50
  • Store below 10 to 30 °C


These agar bases contain peptone, special (23.000 g/l); corn starch (1.000 g/l); sodium chloride (5.000 g/l) and agar (15.000 g/l).


Certifications: In accordance with the harmonazied method of USP/EP/BP/JP.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR