STARRAM 137 Monolithic Refractory Dry Cement, Morgan Advanced Materials
Supplier: Morgan Advanced Materials
95-1000-0001000 695-1000-0001000
11019-826EA 424.29 CAD
11019-826 CA11019-826
STARRAM 137 Monolithic Refractory Dry Cement, Morgan Advanced Materials
Adhesives
This granular, odorless solid cement mixture appears white, gray, or brown in color
It has a melting point of 1705°C (3100°F), and a specific gravity of 2.500 to 3.500 g/cc. When handling, it is recommended that workers wear clothing which minimizes skin contact or exposure. The mixture is stable, however it is considered incompatible with strong acids, bases, or oxidizing agents. Store in a cool and dry environment.