Dextrose Salt Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M102-500G M102-500G
CA95020-314EA 1201.02 CAD
95020-314 CA95020-314
Dextrose Salt Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the enumeration of yeasts and molds in butter and other dairy products.
  • Microbiological culture media
  • 500 g poly bottles