Dextrose Tryptone Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M092-500G M092-500G
CA95020-244EA 1031.38 CAD
95020-244 CA95020-244
Dextrose Tryptone Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the detection and enumeration of mesophilic and thermophilic aerobic organisms in foods.
  • Microbiological culture media
  • 500 g poly bottles