Anaerobic Basal Agar Dehydrated Culture Media, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
M1635-500G M1635-2.5KG M1635-5KG
CA95021-386EA 2010.52 CAD
CA95021-386 CA89224-482 CA89224-484
Anaerobic Basal Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the growth of anaerobic microorganisms, particularly Bacteroides species and other fastidious anaerobes.
  • Microbiological culture media
Packaging: Packaged in 500 g poly bottles.