EC Broth Base, Modified, Dehydrated Culture Media, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
M1285-500G M1285-500G
CA95020-866EA 1048.27 CAD
CA95020-866 95020-866
EC Broth Base, Modified, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the isolation of Escherichia coli O157:H7.
  • Microbiological culture media
  • 500 g poly bottles