Tryptic Soy Broth, Irradiated, Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
1.00800.0500 1.00800.5000 1.00800.5000
CA1.00800.5000EA 3612.9 CAD
CA1.00800.5000 EM1.00800.0500 EM1.00800.5000
Tryptic Soy Broth, Irradiated, Dehydrated Culture Media, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
Casein-peptone soymeal-peptone broth USP for microbiology.
  • Granulation preserves homogeneity
  • Easy to weigh out
  • Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.
Media is granulated.