Samco™ Clicktainer™ Specimen Containers, Thermo Scientific
Supplier: Thermo Scientific
Samco™ Clicktainer™
120ORG53-1000 90WHT53-2002 90WHT53-1001 90WHT53-2001 90ORG53-1001 120WHT53-4000 90WHT53-1000 90WHT53-2000 90WHT53-2004 90WHT53-1002 120ORG53-4000 120WHT53-1000 90ORG53-1000 120WHT53-2000 90WHT53-2003 90GRN53-1000
76019-390CS 494.82 CAD
76019-390 76019-380 76019-374 76019-396 76019-386 76019-376 76019-388 76019-392 76019-370 76019-382 76019-384 76019-406 76019-400 76019-378 76019-402 76019-408
Samco™ Clicktainer™ Specimen Containers, Thermo Scientific
Sample Containers General Purpose Containers
Unique click seal system provides ultimate sample protection and user safety by ensuring correct application the first time.
  • Visual, audible, and tactile indication of correct cap closure
  • Wide 53 mm opening for ease of sample collection and management
  • Leakproof performance
  • 95 kPa compliant at ambient temperature
  • Ideal for use in pneumatic tube systems
  • Excellent clarity with easy to see graduations
Thermo Scientific understands the value of patient samples.
With this innovative closure system, Clicktainer™ Vials are ideally suited for the collection and transport of fluids, solids, and powders. They are perfect for a wide range of customer applications, from the collection of valuable and often hazardous patient samples such as urine, feces, and sputum and their transportation through hospital pneumatic/vacuum transport systems, to their suitability for use with formalin or fixatives for histology applications.
Choose safety first with the Samco™ Clicktainer™ Specimen Vials.
Sample collection needs to be easy. Sample transport needs to be secure. The Samco™ Clicktainer™ Vials have a wide opening and leakproof seal. The 'click' seal system indicates when the cap is properly applied so the sample is secure.
Certifications: CE marked in accordance with IVD Directicve 98/79/EC. Each batch pressure tested to 95kPa in accordance with BS EN 14401. Leak tested in accordance with BS EN 14254.
Caution: Note:  The term 'leakproof' applies to Thermo Scientific Samco™ Clicktainer™ products, 90 ml/53 mm and 120 ml/53 mm, that meet the following criteria: a) a vacuum test that draws 95 kPa of pressure within a sealed chamber with no leakage; b) leak testing in accordance with EN 14254, which involves filling the container with a dye, placing for 2 minutes on a roller mixer followed by one hour inverted in a water bath with no leakage. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Samco™ Clicktainer™ Vials under conditions of their planned applications. Vials are leakproof at ambient temperature and pressure when used with their Clicktainer™ closures.