Muffle Furnaces with Brick Insulation, Models L and LT, Nabertherm
Supplier: NABERTHERM INC.
L-054S3LN3-240 L-054S3ON3-240 L-054T3LN3-240 L-054T3ON3-240 L-094S3LN3-208 L-094S3LN3-240 L-094S3ON3-208 L-094S3ON3-240 L-094T3LN3-208 L-094T3LN3-240 L-094T3ON3-208 L-094T3ON3-240 L-154S3LN3-208 L-154S3LN3-240 L-154S3ON3-208 L-154S3ON3-240 L-154T3LN3-208 L-154T3LN3-240 L-154T3ON3-208 L-154T3ON3-240
76537-580EA 11230.8 CAD
76537-580 76537-582 76537-586 76537-588 76537-592 76537-594 76537-598 76537-600 76537-602 76537-604 76537-606 76537-608 76537-610 76537-612 76537-614 76537-616 76537-618 76537-620 76537-622 76537-624 76537-626 76537-628 76537-630 76537-632
Muffle Furnaces with Brick Insulation, Models L and LT, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
Heating elements on support tubes radiating freely into the furnace chamber provide for particularly short heating times for these muffle furnaces.
  • Tmax 1300 °C
  • Heating from two sides
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Multi-layer insulation with robust lightweight refractory bricks in the furnace chamber
  • Housing made of sheets of textured stainless steel
  • Dual shell housing for low external temperatures and stability
  • Optional fold down door (L models) which can be used as work platform or lift up door (LT
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable