EZwaste® UN/DOT 70 mm Cap Assembly for HPLC Solvent Waste, Carbon Air Filter, Foxx Life Sciences
Supplier: Foxx Life Sciences
330-0B01-OEM 330-0B03-OEM 330-0B05-OEM 330-0B11-OEM 330-0B15-OEM 330-0B16-OEM 76018-608 76018-610 76018-612 76018-614 76018-616 76018-618 76018-620 76018-622
75805-410EA 602.7 CAD
75805-410 75805-412 75805-414 75805-416 75805-418 75805-420 76018-608 76018-610 76018-612 76018-614 76018-616 76018-618 76018-620 76018-622
EZwaste® UN/DOT 70 mm Cap Assembly for HPLC Solvent Waste, Carbon Air Filter, Foxx Life Sciences
Filters Liquid Process Filters
This cap system allows for the safe and effective collection of waste in the laboratory. The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.
  • Affordable solution for HPLC liquid waste disposal
  • VersaCap® technology
  • Carbon exhaust filter traps VOCs
  • Variety of ext.Ø tubing port configurations
  • Hose Barb options for ¹/₄ or ³/₈" int.Ø
The EZwaste® UN/DOT Solvent Waste System is designed to be an affordable solution for containing liquid chromatography waste
Depending on application, carbon exhaust filter life cycle may vary.
Each system is manufactured to safely collect waste from up to two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste System also features a closed-loop design which protects against leaks and spills, improving overall lab safety. The S-70 caps come with a variety of port configurations for ¹/₁₆, ¹/₈, ¹/₄" ext.Ø tubing and hose barbs. All UN/DOT containers are rated for transport for easy waste pickup.