EZwaste® XL 40 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Supplier: VWR International
75805-436 75805-510 75805-512 75805-514 75805-516 75805-518 76018-712 76018-714 76018-716 76018-718 76018-720 76018-722 76018-724 76018-726
75805-436EA 1704.62 CAD
75805-436 75805-510 75805-512 75805-514 75805-516 75805-518 76018-712 76018-714 76018-716 76018-718 76018-720 76018-722 76018-724 76018-726
EZwaste® XL 40 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Filters Liquid Process Filters
This 40 L system allows for the safe and effective collection of waste in the laboratory. The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.
  • Affordable solution for HPLC liquid waste disposal
  • VersaCap® technology
  • Carbon exhaust filter traps VOCs
  • Variety of ext.Ø tubing port configurations
  • Hose Barb option in ¹/₄ or ³/₈"
The EZwaste® Solvent Waste System is designed to be an affordable solution for containing liquid chromatography waste.
Each EZwaste®More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Filter dimensions Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
Height 771 mm (30.4") with Cap; 736 mm (29") without Cap