EZwaste® HD Closed System for HPLC Solvent Waste, 10 L, HDPE Reusable Carboy,Foxx Life Sciences
Supplier: VWR International
75805-494 75805-496 75805-498 75805-500 76018-538 76018-540 76018-542 76018-544 76018-546 76018-548 76018-550 76018-552 76018-554
75805-494EA 1289.55 CAD
75805-494 75805-496 75805-498 75805-500 76018-538 76018-540 76018-542 76018-544 76018-546 76018-548 76018-550 76018-552 76018-554
EZwaste® HD Closed System for HPLC Solvent Waste, 10 L, HDPE Reusable Carboy,Foxx Life Sciences
Filters Liquid Process Filters
This 10 L system allows for the safe and effective collection of waste in the laboratory. The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.
  • Affordable solution for HPLC liquid waste disposal
  • VersaCap® technology
  • Carbon exhaust filter traps VOCs
  • Variety of O.D. tubing port configurations
  • Hose Barb options for ¹/₄ or ³/₈" int.Ø. tubes
The EZwaste® Solvent Waste System is designed to be an affordable solution for containing liquid chromatography waste.
Order Now
SPECIFICATIONS
Environmentally Preferable
Filter dimensions Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
Height 496 mm (19.5") with Cap; 487 mm (19.2") without Cap