EZwaste® HD 2.5 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Supplier: VWR International
332-1411-OEM 332-1412-OEM 332-1413-OEM 332-1418-OEM 332-1419-OEM 332-1420-OEM 332-1421-OEM 332-1426-OEM 332-1427-OEM 332-1428-OEM 332-1429-OEM 332-1430-OEM
76018-638EA 974.44 CAD
76018-638 76018-640 76018-642 76018-644 76018-646 76018-648 76018-650 76018-652 76018-654 76018-656 76018-658 76018-684 76018-686 76018-688
EZwaste® HD 2.5 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Filters Liquid Process Filters
This 2.5 L system allows for the safe and effective collection of waste in the laboratory. The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.
  • Affordable solution for HPLC liquid waste disposal
  • VersaCap® technology
  • Carbon exhaust filter traps VOCs
  • Variety of O.D. tubing port configurations
  • Hose Barb option in ¹/₄ or ³/₈"
The EZwaste® Solvent Waste System is designed to be an affordable solution for containing liquid chromatography waste.
Each EZwaste® Solvent Waste System comes with two 83mm VersaCap® (one open top for waste collection and one closed top for safe transport), fittings, plugs for unused ports, and a chemical exhaust filter. Depending on application, carbon exhaust filter life cycle may vary.
Tube fittings secure waste tubes to carboy, providing the closed system necessary to prevent spills. A 90° adapter is included for maximum storage flexibility. A replaceable sulfur cured EPDM gasket is included for high chemical compatibility against aggressive organic solvents.
Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste System also features a closed-loop design, which protects against leaks and spills, improving overall lab safety.