EZwaste® UN/DOT 13.5 L Closed System for HPLC Solvent Waste, HDPE Disposable Container, Foxx Life Sciences
Supplier: VWR International
75805-444 75805-534 75805-536 75805-538 75805-540 75805-542 76018-738 76018-740 76018-742 76018-744 76018-746 76018-748 76018-750 76018-752
75805-444EA 918.51 CAD
75805-444 75805-534 75805-536 75805-538 75805-540 75805-542 76018-738 76018-740 76018-742 76018-744 76018-746 76018-748 76018-750 76018-752
EZwaste® UN/DOT 13.5 L Closed System for HPLC Solvent Waste, HDPE Disposable Container, Foxx Life Sciences
Filters Liquid Process Filters
This system allows for the safe and effective collection of waste in the laboratory. The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.
  • Traps volatile organic compunds, providing a safe working environment for laboratory technicians
  • Closed-loop design allows for the capture of vapor waste
  • 70 mm VersaCap® included
The EZwaste® UN/DOT Solvent Waste System is designed to be an affordable solution for containing liquid chromatography waste.
Each EZwaste® UN/DOT Solvent Waste System comes with one 70 mm open Cap, fittings, plugs for unused ports, and a chemical exhaust filter. Depending on application, carbon exhaust filter life cycle may vary. All UN/DOT containers are rated for transport for easy waste pickup.
Each system is manufactured to safely collect waste from up to two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste System also features a closed-loop design which protects against leaks and spills, improving overall lab safety.