EZFlow® Syringe Filter, Sample Prep, Hydrophilic PTFE, Foxx Life Sciences
Supplier: Foxx Life Sciences
386-2116-OEM 386-2216-OEM 386-2416-OEM 386-3116-OEM 386-3216-OEM 386-3416-OEM
76018-926PK 218.77 CAD
76018-926 76018-928 76018-930 76018-932 76018-934 76018-936
EZFlow® Syringe Filter, Sample Prep, Hydrophilic PTFE, Foxx Life Sciences
Filters Syringe Filters
Hydrophilic PTFE sample prep syringe filters have excellent chemical resistance, are able to be used with aqueous solutions, and are perfect for use with aggressive solvents and with HPLC sample prep.
  • Excellent chemical resistance
  • Perfect for use with aggressive solvents
  • Able to be used with aqueous solutions
  • Great for HPLC sample prep
EZFlow® 13, 25, and 33 mm sample prep syringe filters are designed for aqueous and organic solution samples. Sample prep syringe filters are used for clarification and particle removal from samples before instrumental analysis such as HPLC, IC, and GC. These syringe filters are available in a variety of filter membrane materials, in 0.2 or 0.45 µm pore sizes. The filter membranes are reinforced with polypropylene housing with a female Luer-lok inlet and a male Luer slip outlet.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.