EZFlow® Syringe Filter, Glass Fiber, Foxx Life Sciences
Supplier: Foxx Life Sciences
37C-2116-OEM 37C-2216-OEM 37C-3116-OEM 37C-3216-OEM
76018-898PK 303.6 CAD
76018-898 76018-900 76018-902 76018-904
EZFlow® Syringe Filter, Glass Fiber, Foxx Life Sciences
Filters Syringe Filters
Glass Fiber syringe filters have super low extractables and fast flow rates.
  • Chemically inert membrane
  • Super low extractables
  • Fast flow rates
  • Resistant to almost all solvents
EZFlow® 13, 25, and 33 mm syringe filters are used for prefiltration, clarification, or fine particulate removal. These syringe filters are available in a variety of filter membrane materials, in 0.2 or 0.45 µm pore sizes. The filter membranes are reinforced with polypropylene housing with a female Luer-lok inlet and a male Luer slip outlet.
They are chemically inert and resistant to almost all solvents.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.