Brilliant Green Agar with Phosphates Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M971I-500G M971I-500G
CA95022-560EA 1313.68 CAD
CA95022-560 95022-560
Brilliant Green Agar with Phosphates Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the detection and enumeration of Salmonellae.
  • Microbiological culture media
  • Packaged in 500 g poly bottles