Brilliant Green Bile Broth, 2%, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M121-500G M121S-500G
CA95020-706EA 932.71 CAD
CA95020-706 CA95020-724
Brilliant Green Bile Broth, 2%, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the detection and confirmation of coliform bacteria in water, waste water, food, milk, and other dairy products.
  • Microbiological culture media