Workstation Dust Hoods, Monashee
Supplier: MONASHEE MANUFACTURING CA
107239-02A 107239-09 107239-48 107239-86
CA11022-156EA 9156.06 CAD
CA11022-156 CA11022-170 CA11022-186 CA11022-222
Workstation Dust Hoods, Monashee
Fume Hoods and Enclosures Ductless Fume Hoods
Workstation dust hoods are used where dust is created such as fluxing, cleaning pulverizer bowls or cleaning screens. These units effectively capture and collect dust while providing uniform airflow, a clean work surface and protecting the operator from contaminants. Designed for industrial and mining applications, our workstation dust hoods have a 10" flange for connecting to existing duct work and are required to be connected to an appropriately sized dust collector (sold separately, contact us for more information).
  • Solid steel construction with powder coating for long lasting finish
  • 10" flange to attach to ducting
  • LED lighting
  • Compressed air hose (air compressor sold separately)
Workstation dust hoods are made of steel and blue powder coated for a long-lasting finish (additional colors available on request). In addition to our standard workstation dust hoods sizes of 4, 5 and 6 ft. custom sizes are also available.