Cupellation Hood
Supplier: Monashee
107239-06 107239-06
CA11022-218EA 2504.55 CAD
CA11022-218 11022-218
Cupellation Hood
Fume Hoods and Enclosures Ductless Fume Hoods
The cupellation hood is designed for the MAS3 furnace and is to be used in conjunction with a furnace cap hood (11022-166). The cupellation hood bolts onto the front of the furnace cap hood and effectively collects lead fumes generated by the hot cupels after cupellation.
  • Designed to fit over the MAS 2 and MAS 3 furnaces
  • Solid steel construction with powder coating for long lasting finish
Cupellation hoods are made of steel and white powder coated for a long-lasting finish (additional colours available on request).