Test Kits, COD, TNT plus®
Supplier: Hach
TNT820 TNT823 TNT821 TNT822 TNT825
CA11014-646PK 185.11 CAD
CA11014-646 CA11016-932 CA11016-948 CA11016-950 CA11016-952 CA11024-890 CA11024-892 75919-242 75919-244 75919-246 75919-248 75919-300 75919-302 75919-304 75919-306 75919-310
Test Kits, COD, TNT plus®
Environmental Test Kits
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are color coded by parameter and range
  • No reagent blank necessary
TNTplus™ works only with DR 6000™ UV-VIS, DR 3900™ Benchtop, and DR 1900™ Portable spectrophotometers.