SHOWA VM Flock-Lined, Natural Rubber, 18 mil, 12 Glove, Showa
Supplier: Showa
VMXL-10 VMS-07 VMM-08
CA32892-087CS 407.67 CAD
CA32892-087 CA32892-088 CA32892-090
SHOWA VM Flock-Lined, Natural Rubber, 18 mil, 12 Glove, Showa
Gloves
The flocked lined chemical resistant gloves provides comfort and flexibility.
  • Outstanding tactile sensitivity, strength, dexterity and elasticity
  • Self flushing tractor-tread pattern provides an excellent grip, wet or dry
  • Glove encourages run off of fluids
These gloves are used in janitorial, food processing, and lab environments. 18 mil thickness provides added protection when working with light chemicals.
Certifications: CE Cat. I marked.