EZFlow® Syringe Filter, Hydrophilic PVDF, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
378-2215-OEM 378-2415-OEM 378-3115-OEM 378-3215-OEM 378-3415-OEM
76018-812PK 454.03 CAD
76018-812 76018-814 76018-816 76018-818 76018-820 76018-882
EZFlow® Syringe Filter, Hydrophilic PVDF, Sterile, Foxx Life Sciences
Filters Syringe Filters
Hydrophilic PVDF syringe filters are great for HPLC sample prep.
  • Hydrophilic
  • Resistant to many solvents
  • Low protein binding
  • Great for HPLC sample prep
EZFlow® 13, 25, and 33 mm sample prep syringe filters are designed for aqueous and organic solution samples. The sample prep syringe filters are used for clarification and particle removal from samples before instrumental analysis such as HPLC, IC, and GC. These syringe filters are available in a variety of filter membrane materials, in 0.2 or 0.45 µm pore sizes. The filter membranes are reinforced with polypropylene housing with a female Luer-lok inlet and a male Luer slip outlet.
They are resistant to many solvents and low protein binding.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.