EZFlow® High Particulate Syringe Filter, PES with Glass Fiber Prefilter, Foxx Life Sciences
Supplier: Foxx Life Sciences
372-2216-OEM 372-3216-OEM
76018-878PK 444.14 CAD
76018-878 76018-880
EZFlow® High Particulate Syringe Filter, PES with Glass Fiber Prefilter, Foxx Life Sciences
Filters Syringe Filters
High particulate PES syringe filters have a 1.0 µm glass fiber prefilter designed for high throughputs and flow rates.
  • Super high flow rates
  • Very low protein binding
  • Very low extractables
  • Perfect for use with media, buffers, and DNA
EZFlow® 25 mm high particulate syringe filters are designed for high throughput filtration of viscous and particle-laden solutions. The syringe filter contains a filter membrane along with a glass fiber prefilter. The filter membrane is available in PES or Hydrophilic PVDF materials, in 0.2 or 0.45 µm pore sizes. The filter membrane is reinforced with a polypropylene housing with a female Luer-Lok inlet and a male Luer Slip outlet.
They also have very low extractables.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.