EZFlow® Syringe Filter, PES, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
371-2115-OEM 371-2215-OEM 371-2415-OEM 371-3115-OEM 371-3215-OEM 371-3415-OEM
76018-866PK 347.81 CAD
76018-866 76018-868 76018-870 76018-872 76018-874 76018-876
EZFlow® Syringe Filter, PES, Sterile, Foxx Life Sciences
Filters Syringe Filters
High performance PES syringe filters have super high flow rates, very low extractables, and are very low protein binding.
  • Super high flow rates
  • Very low protein binding
  • Very low extractables
  • Perfect for use with media, buffers, and DNA
EZFlow® 13, 25, and 33 mm Syringe Filters are used for prefiltration, clarification, sterilization, or fine particulate removal. These syringe filters are available in a variety of filter membrane materials in both 0.2 and 0.45 µm pore sizes. The filter membranes are reinforced with polypropylene housing, with a female Luer-lok inlet and a male Luer slip outlet.
All at an affordable cost.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.