EZFlow® Membrane Disc Filter, 90 mm, 1.0 µm Glass Fiber, Nonsterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
36C-9831-FLS 36C-9832-FLS 36C-9833-FLS
76018-860PK 90.19 CAD
76018-860 76018-862 76018-864
EZFlow® Membrane Disc Filter, 90 mm, 1.0 µm Glass Fiber, Nonsterile, Foxx Life Sciences
Filters Membrane Filters
Glass Fiber (GF) membranes have super low extractables and fast flow rates.
  • Chemically inert membrane
  • Resistant to almost all solvents
  • Sterilized by autoclaving
EZFlow® 90 mm Glass Fiber membrane disc filters are manufactured from Borosilicate glass without binders. These glass fiber disc filters have good solids holding capacity and can be used as pre-filters for heavily contaminated solution samples. The glass fiber membrane disc filters have high flow rates, wet strength, and are available in a 1.00 µm pore size.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.