EZFlow® Membrane Disc Filter, Hydrophobic PTFE, Foxx Life Sciences
Supplier: Foxx Life Sciences
365-2212-OEM 365-2612-OEM 365-2811-OEM 365-3212-OEM 365-3612-OEM 365-3811-OEM
76018-848PK 73.04 CAD
76018-848 76018-850 76018-852 76018-854 76018-856 76018-858
EZFlow® Membrane Disc Filter, Hydrophobic PTFE, Foxx Life Sciences
Filters Membrane Filters
Hydrophobic PTFE membranes have excellent chemical resistance.
  • Excellent chemical resistance
  • Perfect for use with aggressive solvents
  • Great for HPLC sample prep
EZFlow® 25, 47 and 90 mm membrane disc filters are used to retain particles or microorganisms by surface capture. The Hydrophobic PTFE membrane is bonded to a polypropylene support. Due to its extreme chemical compatibility, they are ideal for the filtration of solvents and acids, or other aggressive chemicals. They are offered in 0.22 or 0.45 µm pore sizes.
Certifications: All materialsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable