EZFlow® Membrane Disc Filter, PES, Foxx Life Sciences
Supplier: Foxx Life Sciences
361-3113-OEM 361-3212-OEM 361-3612-OEM 361-3811-OEM 361-2113-OEM 361-2212-OEM 361-2612-OEM 361-2811-OEM
76018-796PK 260.67 CAD
76018-796 76018-798 76018-800 76018-802 76018-804 76018-806 76018-808 76018-810
EZFlow® Membrane Disc Filter, PES, Foxx Life Sciences
Filters Membrane Filters
High performance PES disc filters have super high flow rates.
  • Very low protein binding
  • Very low extractables
  • Perfect for use with media, buffers, and DNA
EZFlow® 13, 25, 47 and 90 mm membrane disc filters are used to retain particles or microorganisms by surface capture. The membrane is offered in 0.22 and 0.45 µm pore sizes.
All at an affordable cost.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.