EZBio® Single Use-Cap Assembly, VersaCap, Polypropylene, Vented with Tube, Foxx Life Sciences
Supplier: Foxx Life Sciences
430-0014-FLS 430-0015-FLS 430-0016-FLS 430-0017-FLS 430-0018-FLS
76018-446CS 2453 CAD
76018-446 76018-448 76018-450 76018-452 76018-454
EZBio® Single Use-Cap Assembly, VersaCap, Polypropylene, Vented with Tube, Foxx Life Sciences
Closures Safety Caps and Safety Waste Caps
Vented EZBio® Single-Use Cap Assemblies are an innovative and cost efficient option for single-use applications
  • Perfect for fluid transfer and storage
  • VersaCap® technology with tubing and vent filters
  • More cost efficient than stainless steel apparatuses
  • USP Class VI components
  • Autoclavable
The EZbio® Single-Use Cap Assembly for bottles with Tubing and Vent Filters are perfect for fluid transfer and storage. Assemblies are made entirely with USP Class VI components. Assemblies take advantage of Foxx’s patented VersaCap® technology, which means there is no machining necessary to produce the product. Perfect for single-use applications and to phase out costly stainless steel apparatuses that are expensive to clean and validate.
They are autoclavable and made entirely with USP Class VI components.
Certifications: All materials comply with USP Class VI standards.