Volumetric Flasks
Supplier: Wards
CH0441A CH0441B CH0441C CH0441E CH0441F CH0441G
470228-950EA 16.25 CAD
470228-950 470228-952 470228-954 470228-956 470228-958 470228-960
Volumetric Flasks
Flasks Volumetric Flasks
Class A Flasks.
  • Blue graduations
  • Glass stopper
Perfect for everyday use in the laboratory, these borosilicate glass flasks are marked with blue graduations for easy reading and fitted with a solid glass stopper. In compliance with ASTM-E288 standard.